The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016131.4 | NP_057215.3 | 4 - 5 | 1168 | 125 |
GenBank mRNA | AF086917.1 | - | 4 - 5 | 513 | 125 |
AF106681.1 | - | 4 - 5 | 509 | 125 | |
AF297660.1 | - | 4 - 5 | 419 | 125 | |
AF498945.1 | - | 4 - 5 | 419 | 125 | |
AK023223.1 | - | 4 - 5 | 918 | 125 | |
BC000896.1 | - | 4 - 5 | 494 | 125 | |
CR457303.1 | - | 4 - 5 | 419 | 125 |