The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308207.1 | NP_001295136.1 | 4 - 5 | 1065 | 92 |
NM_001308208.1 | NP_001295137.1 | 3 - 4 | 718 | 92 | |
NM_025251.2 | NP_079527.1 | 2 - 3 | 591 | 92 | |
XM_011517308.1 | XP_011515610.1 | 3 - 4 | 872 | 92 | |
XM_011517309.1 | XP_011515611.1 | 3 - 4 | 776 | 92 | |
XM_017013870.1 | XP_016869359.1 | 4 - 5 | 1944 | 92 | |
XM_017013871.1 | XP_016869360.1 | 3 - 4 | 1003 | 92 | |
GenBank mRNA | AB051475.1 | - | 4 - 5 | 1069 | 92 |
AK296353.1 | - | 3 - 4 | 715 | 92 |