Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKD1
Entrez Gene ID
85407
Gene Name
naked cuticle homolog 1
Gene Aliases
Naked1
Chromosome Location
Chr.16: 50548330 - 50649249 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033119.4 NP_149110.1 5 - 6 590 103
XM_011523399.2 XP_011521701.1 8 - 9 901 103
XM_011523400.2 XP_011521702.1 3 - 4 192 103
XM_017023804.1 XP_016879293.1 7 - 8 2120 103
GenBank mRNA AB062886.1 - 5 - 6 371 103
AF289584.1 - 5 - 6 642 103
AF358135.1 - 5 - 6 373 103
AK314930.1 - 5 - 6 524 103
AY061883.1 - 5 - 6 524 103
BC051288.1 - 5 - 6 590 103

View Full Product Details