The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033119.4 | NP_149110.1 | 9 - 10 | 1046 | 104 |
XM_011523399.2 | XP_011521701.1 | 12 - 13 | 1357 | 104 | |
XM_011523400.2 | XP_011521702.1 | 7 - 8 | 648 | 104 | |
XM_017023804.1 | XP_016879293.1 | 11 - 12 | 2576 | 104 | |
GenBank mRNA | AB062886.1 | - | 9 - 10 | 827 | 104 |
AF289584.1 | - | 9 - 10 | 1098 | 104 | |
AF358135.1 | - | 9 - 10 | 829 | 104 | |
AK314930.1 | - | 9 - 10 | 980 | 104 | |
AY061883.1 | - | 9 - 10 | 980 | 104 | |
BC051288.1 | - | 9 - 10 | 1046 | 104 |