The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109754.2 | NP_001103224.1 | 32 - 33 | 6546 | 84 |
NM_001206971.1 | NP_001193900.1 | 29 - 30 | 5607 | 84 | |
NM_001206972.1 | NP_001193901.1 | 29 - 30 | 5607 | 84 | |
NM_002837.4 | NP_002828.3 | 30 - 31 | 5877 | 84 | |
XM_006719528.1 | XP_006719591.1 | 32 - 33 | 6546 | 84 | |
XM_006719529.3 | XP_006719592.1 | 30 - 31 | 6373 | 84 | |
XM_011538614.1 | XP_011536916.1 | 31 - 32 | 6282 | 84 | |
XM_017019723.1 | XP_016875212.1 | 31 - 32 | 6282 | 84 | |
XM_017019724.1 | XP_016875213.1 | 24 - 25 | 4321 | 84 | |
GenBank mRNA | BC101679.1 | - | 30 - 31 | 5876 | 84 |
BC113463.1 | - | 30 - 31 | 5876 | 84 | |
BC143356.1 | - | 29 - 30 | 5606 | 84 | |
BC143360.1 | - | 29 - 30 | 5606 | 84 | |
BE042873.1 | - | 2 - 3 | 84 | ||
BX648245.1 | - | 32 - 33 | 6529 | 84 | |
BX648771.1 | - | 29 - 30 | 5725 | 84 | |
CR749546.1 | - | 31 - 32 | 6262 | 84 | |
X54131.1 | - | 30 - 31 | 5877 | 84 |