The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109754.2 | NP_001103224.1 | 33 - 34 | 6669 | 171 |
NM_001206971.1 | NP_001193900.1 | 30 - 31 | 5730 | 171 | |
NM_001206972.1 | NP_001193901.1 | 30 - 31 | 5730 | 171 | |
NM_002837.4 | NP_002828.3 | 31 - 32 | 6000 | 171 | |
XM_017019723.1 | XP_016875212.1 | 32 - 33 | 6405 | 171 | |
GenBank mRNA | BC101679.1 | - | 31 - 32 | 5999 | 171 |
BC113463.1 | - | 31 - 32 | 5999 | 171 | |
BC143356.1 | - | 30 - 31 | 5729 | 171 | |
BC143360.1 | - | 30 - 31 | 5729 | 171 | |
BE042873.1 | - | 1 - 2 | 171 | ||
BX648245.1 | - | 33 - 34 | 6652 | 171 | |
BX648771.1 | - | 30 - 31 | 5848 | 171 | |
CR749546.1 | - | 32 - 33 | 6385 | 171 |