Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAMD3
Entrez Gene ID
65983
Gene Name
GRAM domain containing 3
Gene Aliases
NS3TP2
Chromosome Location
Chr.5: 126360096 - 126494377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146319.1 NP_001139791.1 13 - 14 1531 93
NM_001146320.1 NP_001139792.1 12 - 13 1568 93
NM_001146321.1 NP_001139793.1 14 - 15 1723 93
NM_001146322.1 NP_001139794.1 13 - 14 1342 93
NM_023927.2 NP_076416.2 13 - 14 1680 93
XM_005272057.3 XP_005272114.1 13 - 14 1421 93
XM_005272058.3 XP_005272115.1 13 - 14 1418 93
XM_005272059.4 XP_005272116.1 14 - 15 1811 93
XM_005272060.3 XP_005272117.1 13 - 14 1508 93
XM_005272061.3 XP_005272118.1 13 - 14 1503 93
XM_005272062.3 XP_005272119.1 13 - 14 1469 93
XM_005272063.3 XP_005272120.1 13 - 14 1615 93
XM_005272064.3 XP_005272121.1 14 - 15 1528 93
XM_011543591.2 XP_011541893.1 14 - 15 2399 93
XM_011543592.2 XP_011541894.1 14 - 15 2392 93
XM_011543593.2 XP_011541895.1 13 - 14 1508 93
XM_011543594.2 XP_011541896.1 13 - 14 1280 93
XM_011543595.2 XP_011541897.1 12 - 13 1275 93
XM_017009781.1 XP_016865270.1 14 - 15 1286 93
XM_017009782.1 XP_016865271.1 14 - 15 1525 93
XM_017009784.1 XP_016865273.1 12 - 13 1327 93
XM_017009785.1 XP_016865274.1 12 - 13 1377 93
GenBank mRNA AK024966.1 - 13 - 14 1448 93
AK225829.1 - 13 - 14 1448 93
AK293307.1 - 12 - 13 1273 93
AK294656.1 - 13 - 14 1329 93
AK296267.1 - 14 - 15 1428 93
AK298009.1 - 13 - 14 1265 93
AK300154.1 - 13 - 14 1476 93
AK303857.1 - 13 - 14 1282 93
AK316288.1 - 13 - 14 1495 93
AY116970.1 - 13 - 14 1250 93
BC008590.1 - 13 - 14 1413 93

View Full Product Details