The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025185.3 | NP_079461.2 | 11 - 12 | 1774 | 144 |
XM_005257203.4 | XP_005257260.1 | 12 - 13 | 2001 | 144 | |
XM_006721806.3 | XP_006721869.1 | 13 - 14 | 2223 | 144 | |
XM_006721807.3 | XP_006721870.1 | 13 - 14 | 2223 | 144 | |
XM_006721810.3 | XP_006721873.1 | 13 - 14 | 2223 | 144 | |
XM_006721811.3 | XP_006721874.1 | 13 - 14 | 2223 | 144 | |
XM_011524597.2 | XP_011522899.1 | 13 - 14 | 2223 | 144 | |
XM_011524598.2 | XP_011522900.1 | 13 - 14 | 2223 | 144 | |
XM_011524599.1 | XP_011522901.1 | 13 - 14 | 2049 | 144 | |
XM_011524600.2 | XP_011522902.1 | 12 - 13 | 2112 | 144 | |
XM_011524601.2 | XP_011522903.1 | 13 - 14 | 2112 | 144 | |
XM_011524602.2 | XP_011522904.1 | 12 - 13 | 2001 | 144 | |
XM_017024429.1 | XP_016879918.1 | 13 - 14 | 2049 | 144 | |
XM_017024430.1 | XP_016879919.1 | 9 - 10 | 5390 | 144 | |
XM_017024431.1 | XP_016879920.1 | 13 - 14 | 2223 | 144 | |
XM_017024432.1 | XP_016879921.1 | 13 - 14 | 2223 | 144 | |
GenBank mRNA | AB046856.2 | - | 7 - 8 | 1375 | 144 |