The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144648.1 | NP_653249.1 | 9 - 10 | 1128 | 141 |
XM_017011749.1 | XP_016867238.1 | 9 - 10 | 2571 | 141 | |
GenBank mRNA | AK057348.1 | - | 9 - 10 | 1128 | 141 |
BC104897.1 | - | 9 - 10 | 1181 | 141 | |
BC104899.1 | - | 9 - 10 | 1181 | 141 |