The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002271.4 | NP_002262.3 | 25 - 26 | 2846 | 66 |
XM_005254052.3 | XP_005254109.1 | 24 - 25 | 2756 | 66 | |
XM_005254053.3 | XP_005254110.1 | 24 - 25 | 2773 | 66 | |
XM_011521087.2 | XP_011519389.1 | 26 - 27 | 2937 | 66 | |
XM_011521088.2 | XP_011519390.1 | 26 - 27 | 3003 | 66 | |
XM_011521089.2 | XP_011519391.1 | 27 - 28 | 3060 | 66 | |
XM_011521090.2 | XP_011519392.1 | 25 - 26 | 2896 | 66 | |
XM_017020562.1 | XP_016876051.1 | 25 - 26 | 2877 | 66 | |
GenBank mRNA | AK125031.1 | - | 12 - 13 | 2325 | 66 |
AK225286.1 | - | 22 - 23 | 2631 | 66 | |
AK302812.1 | - | 22 - 23 | 2618 | 66 | |
AK303494.1 | - | 20 - 21 | 2259 | 66 | |
BC000947.2 | - | 3 - 4 | 252 | 66 | |
BC001497.2 | - | 22 - 23 | 2614 | 66 | |
BC019309.1 | - | 22 - 23 | 2614 | 66 | |
BC045640.1 | - | 25 - 26 | 2827 | 66 | |
BX648222.1 | - | 22 - 23 | 2676 | 66 | |
U72761.1 | - | 22 - 23 | 2612 | 66 | |
Y08890.1 | - | 25 - 26 | 2848 | 66 |