Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARHGEF10L
- Entrez Gene ID
-
55160
- Gene Name
- Rho guanine nucleotide exchange factor 10 like
- Gene Aliases
-
GrinchGEF
- Chromosome Location
-
Chr.1: 17520197 - 17697875 on Build GRCh38
View Full Product Details