The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030019.1 | NP_001025190.1 | 2 - 3 | 365 | 139 |
NM_152782.3 | NP_689995.3 | 3 - 4 | 281 | 139 | |
XM_017011930.1 | XP_016867419.1 | 3 - 4 | 271 | 139 | |
GenBank mRNA | AF429967.1 | - | 2 - 3 | 365 | 139 |
BC026189.2 | - | 3 - 4 | 269 | 139 | |
BG772576.1 | - | 3 - 4 | 286 | 139 | |
BI825617.1 | - | 2 - 3 | 344 | 139 |