The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167890.1 | NP_001161362.1 | 6 - 7 | 936 | 108 |
NM_015507.3 | NP_056322.2 | 6 - 7 | 936 | 108 | |
GenBank mRNA | AF186084.1 | - | 6 - 7 | 794 | 108 |
AF193055.1 | - | 6 - 7 | 861 | 108 | |
AJ245671.1 | - | 6 - 7 | 889 | 108 | |
AK075214.1 | - | 6 - 7 | 906 | 108 | |
AK300137.1 | - | 6 - 7 | 672 | 108 | |
AK315018.1 | - | 6 - 7 | 774 | 108 | |
AY358333.1 | - | 6 - 7 | 783 | 108 | |
BC038587.1 | - | 6 - 7 | 745 | 108 |