Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AZIN1
Entrez Gene ID
51582
Gene Name
antizyme inhibitor 1
Gene Aliases
AZI, AZIA1, OAZI, OAZIN, ODC1L
Chromosome Location
Chr.8: 102826302 - 102864200 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301668.1 NP_001288597.1 9 - 10 1524 101
NM_015878.5 NP_056962.2 10 - 11 1670 101
NM_148174.3 NP_680479.1 9 - 10 1524 101
XM_011517126.1 XP_011515428.1 10 - 11 1657 101
XM_011517127.1 XP_011515429.1 10 - 11 1648 101
XM_011517128.2 XP_011515430.1 9 - 10 1668 101
XM_017013565.1 XP_016869054.1 10 - 11 1787 101
GenBank mRNA AF070634.1 - 1 - 2 83 101
AK095493.1 - 9 - 10 1452 101
AK290516.1 - 10 - 11 1402 101
BC013420.2 - 10 - 11 1609 101
BC019279.1 - 9 - 10 1471 101
BC042404.1 - 9 - 10 1495 101
BT006858.1 - 7 - 8 897 101
CR456745.1 - 7 - 8 897 101
D88674.1 - 10 - 11 1606 101

View Full Product Details