The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301668.1 | NP_001288597.1 | 4 - 5 | 905 | 67 |
NM_015878.5 | NP_056962.2 | 5 - 6 | 1051 | 67 | |
NM_148174.3 | NP_680479.1 | 4 - 5 | 905 | 67 | |
XM_005250969.1 | XP_005251026.1 | 4 - 5 | 866 | 67 | |
XM_011517126.1 | XP_011515428.1 | 5 - 6 | 1038 | 67 | |
XM_011517127.1 | XP_011515429.1 | 5 - 6 | 1029 | 67 | |
XM_011517128.2 | XP_011515430.1 | 4 - 5 | 898 | 67 | |
XM_017013563.1 | XP_016869052.1 | 5 - 6 | 1017 | 67 | |
XM_017013564.1 | XP_016869053.1 | 5 - 6 | 1029 | 67 | |
XM_017013565.1 | XP_016869054.1 | 5 - 6 | 1017 | 67 | |
XM_017013566.1 | XP_016869055.1 | 4 - 5 | 898 | 67 | |
GenBank mRNA | AK095493.1 | - | 4 - 5 | 833 | 67 |
AK290516.1 | - | 5 - 6 | 783 | 67 | |
BC013420.2 | - | 5 - 6 | 990 | 67 | |
BC019279.1 | - | 4 - 5 | 852 | 67 | |
BC042404.1 | - | 4 - 5 | 876 | 67 | |
BT006858.1 | - | 2 - 3 | 278 | 67 | |
CR456745.1 | - | 2 - 3 | 278 | 67 | |
D88674.1 | - | 5 - 6 | 987 | 67 |