The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152924.4 | NP_690888.1 | 1 - 2 | 411 | 69 |
GenBank mRNA | AF546700.1 | - | 1 - 2 | 143 | 69 |
AK222841.1 | - | 1 - 2 | 83 | 69 | |
AK223083.1 | - | 1 - 2 | 81 | 69 | |
AK290340.1 | - | 1 - 2 | 83 | 69 | |
BC090052.1 | - | 1 - 2 | 145 | 69 | |
BI464471.1 | - | 1 - 2 | 176 | 69 | |
DC397201.1 | - | 1 - 2 | 412 | 69 | |
X12433.1 | - | 1 - 2 | 66 | 69 |