The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007011.7 | NP_008942.3 | 9 - 10 | 1458 | 57 |
NM_152924.4 | NP_690888.1 | 5 - 6 | 1060 | 57 | |
XM_005254832.1 | XP_005254889.1 | 8 - 9 | 1096 | 57 | |
XM_005254833.1 | XP_005254890.1 | 5 - 6 | 704 | 57 | |
XM_011521165.1 | XP_011519467.1 | 6 - 7 | 909 | 57 | |
GenBank mRNA | AF546700.1 | - | 5 - 6 | 792 | 57 |
AK222841.1 | - | 5 - 6 | 732 | 57 | |
AK223083.1 | - | 5 - 6 | 730 | 57 | |
AK290340.1 | - | 5 - 6 | 732 | 57 | |
BC019248.1 | - | 9 - 10 | 1188 | 57 | |
BC090052.1 | - | 5 - 6 | 794 | 57 | |
X12433.1 | - | 5 - 6 | 715 | 57 |