The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001855.4 | NP_001846.3 | 11 - 12 | 1996 | 63 |
XM_011518214.2 | XP_011516516.1 | 12 - 13 | 1991 | 63 | |
GenBank mRNA | AK095885.1 | - | 11 - 12 | 1712 | 63 |
D21230.1 | - | 11 - 12 | 1740 | 63 | |
L25286.1 | - | 11 - 12 | 1736 | 63 |