The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001855.4 | NP_001846.3 | 18 - 19 | 2566 | 68 |
XM_011518214.2 | XP_011516516.1 | 19 - 20 | 2561 | 68 | |
GenBank mRNA | L01697.1 | - | 8 - 9 | 588 | 68 |
L25286.1 | - | 18 - 19 | 2306 | 68 |