The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034837.2 | NP_001030009.1 | 6 - 7 | 1004 | 156 |
NM_001034838.2 | NP_001030010.1 | 5 - 6 | 851 | 156 | |
NM_001278339.1 | NP_001265268.1 | 5 - 6 | 1046 | 156 | |
NM_001278340.1 | NP_001265269.1 | 6 - 7 | 1133 | 156 | |
NM_014592.3 | NP_055407.1 | 5 - 6 | 971 | 156 | |
XM_017009407.1 | XP_016864896.1 | 6 - 7 | 975 | 156 | |
GenBank mRNA | AF199597.1 | - | 5 - 6 | 434 | 156 |
AK074390.1 | - | 5 - 6 | 567 | 156 | |
AK301775.1 | - | 5 - 6 | 626 | 156 | |
AY170821.1 | - | 5 - 6 | 647 | 156 | |
AY780424.1 | - | 6 - 7 | 809 | 156 | |
BC035032.1 | - | 5 - 6 | 976 | 156 | |
BC050375.1 | - | 5 - 6 | 738 | 156 | |
DQ148476.1 | - | 5 - 6 | 461 | 156 | |
DQ148477.1 | - | 5 - 6 | 434 | 156 | |
DQ148478.1 | - | 6 - 7 | 467 | 156 | |
DQ148479.1 | - | 5 - 6 | 509 | 156 |