Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF10L
Entrez Gene ID
55160
Gene Name
Rho guanine nucleotide exchange factor 10 like
Gene Aliases
GrinchGEF
Chromosome Location
Chr.1: 17520197 - 17697875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK001383.1 - 15 - 16 1878 84

View Full Product Details