The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001855.4 | NP_001846.3 | 36 - 37 | 3750 | 110 |
XM_011518214.2 | XP_011516516.1 | 37 - 38 | 3745 | 110 | |
GenBank mRNA | L01697.1 | - | 26 - 27 | 1772 | 110 |
L25286.1 | - | 36 - 37 | 3490 | 110 |