Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
Chromosome Location
Chr.3: 15450133 - 15521751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005677.3 NP_005668.2 4 - 5 493 92
NM_080538.2 NP_536799.1 4 - 5 622 92
NM_080539.3 NP_536800.2 3 - 4 391 92
GenBank mRNA AF057036.1 - 4 - 5 493 92
AJ225895.1 - 4 - 5 486 92
AK128401.1 - 4 - 5 622 92
AK289688.1 - 4 - 5 512 92
AY150334.1 - 4 - 5 470 92
AY150335.1 - 4 - 5 470 92
AY150336.1 - 4 - 5 470 92
AY150337.1 - 4 - 5 470 92
AY150338.1 - 4 - 5 470 92
AY150339.1 - 4 - 5 470 92
BC074828.2 - 4 - 5 397 92
BC074829.2 - 4 - 5 397 92
BX500613.1 - 3 - 4 378 92

View Full Product Details