The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004638.3 | NP_004629.3 | 27 - 28 | 6222 | 81 |
NM_080686.2 | NP_542417.2 | 27 - 28 | 6185 | 81 | |
GenBank mRNA | AB210034.1 | - | 27 - 28 | 4299 | 81 |
AK297210.1 | - | 14 - 15 | 3701 | 81 | |
AK298584.1 | - | 11 - 12 | 1522 | 81 | |
BC030127.1 | - | 11 - 12 | 1555 | 81 | |
BC042295.1 | - | 27 - 28 | 6148 | 81 | |
BC060668.1 | - | 27 - 28 | 6109 | 81 | |
BG423867.1 | - | 4 - 5 | 555 | 81 | |
CR749245.1 | - | 12 - 13 | 3327 | 81 | |
M33509.1 | - | 28 - 29 | 5992 | 81 |