The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004638.3 | NP_004629.3 | 7 - 8 | 1041 | 59 |
NM_080686.2 | NP_542417.2 | 7 - 8 | 1004 | 59 | |
XM_017011274.1 | XP_016866763.1 | 6 - 7 | 756 | 59 | |
GenBank mRNA | AB210034.1 | - | 7 - 8 | 990 | 59 |
AK302756.1 | - | 7 - 8 | 965 | 59 | |
AK309774.1 | - | 7 - 8 | 965 | 59 | |
BC042295.1 | - | 7 - 8 | 967 | 59 | |
BC060668.1 | - | 7 - 8 | 928 | 59 | |
M33509.1 | - | 7 - 8 | 856 | 59 |