The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015507.3 | NP_056322.2 | 9 - 10 | 1465 | 68 |
GenBank mRNA | AF186084.1 | - | 9 - 10 | 1323 | 68 |
AF193055.1 | - | 9 - 10 | 1390 | 68 | |
AK315018.1 | - | 9 - 10 | 1303 | 68 | |
AL117610.1 | - | 9 - 10 | 1195 | 68 | |
AY358333.1 | - | 9 - 10 | 1311 | 68 | |
BC038587.1 | - | 9 - 10 | 1274 | 68 |