The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002831.5 | NP_002822.2 | 8 - 9 | 1172 | 72 |
NM_080548.4 | NP_536858.1 | 8 - 9 | 1085 | 72 | |
NM_080549.3 | NP_536859.1 | 8 - 9 | 1172 | 72 | |
XM_006718994.1 | XP_006719057.1 | 8 - 9 | 1229 | 72 | |
XM_011520988.1 | XP_011519290.1 | 8 - 9 | 1086 | 72 | |
GenBank mRNA | AF178946.1 | - | 8 - 9 | 954 | 72 |
AK223629.1 | - | 8 - 9 | 1067 | 72 | |
AK290421.1 | - | 8 - 9 | 1055 | 72 | |
AK298470.1 | - | 7 - 8 | 1338 | 72 | |
AK300167.1 | - | 7 - 8 | 929 | 72 | |
AK308230.1 | - | 8 - 9 | 1121 | 72 | |
AK313379.1 | - | 8 - 9 | 1085 | 72 | |
BC002523.2 | - | 8 - 9 | 1071 | 72 | |
BC007667.2 | - | 8 - 9 | 1071 | 72 | |
BM006699.1 | - | 2 - 3 | 183 | 72 | |
BT007390.1 | - | 7 - 8 | 936 | 72 | |
M74903.1 | - | 8 - 9 | 1013 | 72 | |
M77273.1 | - | 8 - 9 | 1074 | 72 |