The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152755.1 | NP_689968.1 | 3 - 4 | 439 | 56 |
GenBank mRNA | AK075537.1 | - | 3 - 4 | 393 | 56 |
AY359100.1 | - | 3 - 4 | 443 | 56 | |
BC019903.1 | - | 3 - 4 | 369 | 56 | |
BC032339.1 | - | 3 - 4 | 439 | 56 |