The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282910.1 | NP_001269839.1 | 3 - 4 | 397 | 75 |
NM_001282911.1 | NP_001269840.1 | 4 - 5 | 478 | 75 | |
GenBank mRNA | AK304014.1 | - | 3 - 4 | 332 | 75 |
AK316432.1 | - | 4 - 5 | 413 | 75 |