The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024744.15 | NP_079020.13 | 1 - 2 | 885 | 135 |
XM_005246858.3 | XP_005246915.1 | 2 - 3 | 113 | 135 | |
XM_017004961.1 | XP_016860450.1 | 1 - 2 | 502 | 135 | |
GenBank mRNA | AB053309.1 | - | 1 - 2 | 368 | 135 |