The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243193.1 | NP_001230122.1 | 4 - 5 | 493 | 68 |
NM_016583.3 | NP_057667.1 | 4 - 5 | 493 | 68 | |
NM_130852.2 | NP_570913.1 | 4 - 5 | 493 | 68 | |
GenBank mRNA | AB024937.1 | - | 4 - 5 | 490 | 68 |
AF158745.1 | - | 4 - 5 | 484 | 68 | |
AF172993.1 | - | 4 - 5 | 470 | 68 | |
AF417256.1 | - | 4 - 5 | 493 | 68 | |
AF417257.1 | - | 4 - 5 | 493 | 68 | |
AF439448.1 | - | 4 - 5 | 477 | 68 | |
AK292778.1 | - | 4 - 5 | 492 | 68 | |
AY359020.1 | - | 4 - 5 | 494 | 68 | |
BC012549.1 | - | 4 - 5 | 497 | 68 | |
DQ846869.1 | - | 2 - 3 | 381 | 68 |