Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YTHDC1
Entrez Gene ID
91746
Gene Name
YTH domain containing 1
Gene Aliases
YT521, YT521-B
Chromosome Location
Chr.4: 68310387 - 68350106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031732.2 NP_001026902.1 3 - 4 809 75
NM_133370.2 NP_588611.2 3 - 4 809 75
XM_005265706.2 XP_005265763.1 3 - 4 809 75
XM_005265707.2 XP_005265764.1 3 - 4 943 75
XM_005265708.2 XP_005265765.1 3 - 4 809 75
XM_017008832.1 XP_016864321.1 3 - 4 943 75
XM_017008833.1 XP_016864322.1 3 - 4 943 75
GenBank mRNA AK056430.1 - 3 - 4 794 75
AK098515.1 - 1 - 2 149 75
AL832559.1 - 1 - 2 328 75
BC041119.1 - 3 - 4 739 75
BC053863.1 - 3 - 4 474 75

View Full Product Details