The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100607.2 | NP_001094077.1 | 1 - 2 | 83 | 125 |
GenBank mRNA | BC022261.1 | - | 1 - 2 | 83 | 125 |
BX248011.1 | - | 1 - 2 | 70 | 125 |