Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QRSL1
Entrez Gene ID
55278
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Gene Aliases
GatA
Chromosome Location
Chr.6: 106629566 - 106668417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011535924.2 XP_011534226.1 1 - 2 128 110

View Full Product Details