Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOXHD1
- Entrez Gene ID
-
125336
- Gene Name
- lipoxygenase homology domains 1
- Gene Aliases
-
DFNB77, LH2D1
- Chromosome Location
-
Chr.18: 46476961 - 46657115 on Build GRCh38
View Full Product Details