The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144612.6 | NP_653213.6 | 14 - 15 | 1967 | 62 |
XM_011525804.2 | XP_011524106.1 | 2 - 3 | 201 | 62 | |
XM_017025540.1 | XP_016881029.1 | 6 - 7 | 997 | 62 | |
XM_017025541.1 | XP_016881030.1 | 7 - 8 | 1084 | 62 | |
XM_017025542.1 | XP_016881031.1 | 7 - 8 | 1084 | 62 | |
XM_017025543.1 | XP_016881032.1 | 7 - 8 | 1084 | 62 | |
XM_017025544.1 | XP_016881033.1 | 7 - 8 | 1084 | 62 | |
XM_017025545.1 | XP_016881034.1 | 7 - 8 | 1084 | 62 | |
XM_017025546.1 | XP_016881035.1 | 7 - 8 | 1084 | 62 | |
XM_017025547.1 | XP_016881036.1 | 7 - 8 | 1084 | 62 | |
XM_017025548.1 | XP_016881037.1 | 14 - 15 | 2049 | 62 | |
GenBank mRNA | AK127869.1 | - | 7 - 8 | 1280 | 62 |