The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139278.2 | NP_644807.1 | 3 - 4 | 636 | 64 |
GenBank mRNA | AF467956.1 | - | 3 - 4 | 557 | 64 |
AJ487518.1 | - | 3 - 4 | 766 | 64 | |
AJ487961.1 | - | 3 - 4 | 636 | 64 | |
AY358116.1 | - | 3 - 4 | 374 | 64 |