The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182765.4 | NP_877497.3 | 8 - 9 | 979 | 107 |
NR_104291.1 | 8 - 9 | 979 | 107 | ||
XM_017015750.1 | XP_016871239.1 | 8 - 9 | 829 | 107 | |
XM_017015751.1 | XP_016871240.1 | 8 - 9 | 925 | 107 | |
GenBank mRNA | AK122629.1 | - | 4 - 5 | 270 | 107 |
BC040187.1 | - | 8 - 9 | 922 | 107 | |
BX537856.1 | - | 8 - 9 | 743 | 107 |