The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031848.2 | NP_114036.1 | 1 - 1 | 2517 | 83 |
GenBank mRNA | AF152484.1 | - | 1 - 1 | 2347 | 83 |