Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
Chromosome Location
Chr.5: 140821776 - 141012344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031501.1 NP_113689.1 1 - 1 2339 86
GenBank mRNA AF152483.1 - 1 - 1 2339 86

View Full Product Details