The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281739.2 | NP_001268668.1 | 12 - 13 | 1581 | 86 |
GenBank mRNA | AB084087.1 | - | 12 - 13 | 1441 | 86 |
AK128053.1 | - | 12 - 13 | 1560 | 86 |