The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167941.1 | NP_001161413.1 | 4 - 5 | 607 | 129 |
NM_024908.3 | NP_079184.2 | 4 - 5 | 717 | 129 | |
XM_005254680.3 | XP_005254737.1 | 4 - 5 | 680 | 129 | |
GenBank mRNA | AK023035.1 | - | 4 - 5 | 622 | 129 |
AK290933.1 | - | 4 - 5 | 639 | 129 | |
BC025247.1 | - | 4 - 5 | 648 | 129 | |
BC051855.1 | - | 4 - 5 | 620 | 129 | |
EF011624.1 | - | 4 - 5 | 620 | 129 |