The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167941.1 | NP_001161413.1 | 7 - 8 | 893 | 114 |
NM_024908.3 | NP_079184.2 | 7 - 8 | 1003 | 114 | |
XM_005254680.3 | XP_005254737.1 | 7 - 8 | 966 | 114 | |
GenBank mRNA | AK023035.1 | - | 7 - 8 | 908 | 114 |
AK290933.1 | - | 7 - 8 | 925 | 114 | |
BC025247.1 | - | 7 - 8 | 934 | 114 | |
BC051855.1 | - | 7 - 8 | 906 | 114 | |
EF011624.1 | - | 7 - 8 | 906 | 114 |