The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244949.1 | NP_001231878.1 | 19 - 20 | 2337 | 62 |
NM_020918.5 | NP_065969.3 | 19 - 20 | 2333 | 62 | |
XM_005269998.1 | XP_005270055.1 | 19 - 20 | 2478 | 62 | |
GenBank mRNA | AB046780.1 | - | 13 - 14 | 1625 | 62 |
AK172782.1 | - | 7 - 8 | 890 | 62 | |
AK172799.1 | - | 17 - 18 | 2166 | 62 | |
AL598680.1 | - | 3 - 4 | 244 | 62 | |
AL833061.2 | - | 19 - 20 | 2319 | 62 | |
AL833093.2 | - | 19 - 20 | 2322 | 62 | |
BC030783.1 | - | 19 - 20 | 2301 | 62 | |
BX647451.1 | - | 19 - 20 | 2301 | 62 |