The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000666.2 | NP_000657.1 | 7 - 8 | 847 | 79 |
NM_001198895.1 | NP_001185824.1 | 7 - 8 | 842 | 79 | |
NM_001198897.1 | NP_001185826.1 | 7 - 8 | 847 | 79 | |
NM_001198898.1 | NP_001185827.1 | 6 - 7 | 742 | 79 | |
GenBank mRNA | AK298274.1 | - | 7 - 8 | 596 | 79 |
BC000545.2 | - | 7 - 8 | 591 | 79 | |
BC003023.1 | - | 7 - 8 | 584 | 79 | |
BC014112.2 | - | 7 - 8 | 567 | 79 | |
CD014026.1 | - | 7 - 8 | 688 | 79 | |
CD014027.1 | - | 6 - 7 | 583 | 79 | |
D14524.1 | - | 7 - 8 | 581 | 79 | |
D16307.1 | - | 7 - 8 | 579 | 79 | |
EU794586.1 | - | 7 - 8 | 567 | 79 | |
KU177890.1 | - | 6 - 7 | 523 | 79 | |
L07548.1 | - | 7 - 8 | 584 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316331.1 | NP_001303260.1 | 9 - 10 | 1048 | 79 |
GenBank mRNA | AK298086.1 | - | 9 - 10 | 1024 | 79 |