Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBR4
Entrez Gene ID
23352
Gene Name
ubiquitin protein ligase E3 component n-recognin 4
Gene Aliases
RBAF600, ZUBR1, p600
Chromosome Location
Chr.1: 19074506 - 19210334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020765.2 NP_065816.2 81 - 82 12107 88
XM_011541108.2 XP_011539410.1 83 - 84 12342 88
XM_011541109.2 XP_011539411.1 83 - 84 12339 88
XM_011541110.2 XP_011539412.1 83 - 84 12339 88
XM_011541111.2 XP_011539413.1 83 - 84 12339 88
XM_011541112.2 XP_011539414.1 83 - 84 12327 88
XM_011541113.2 XP_011539415.1 83 - 84 12324 88
XM_011541114.2 XP_011539416.1 83 - 84 12324 88
XM_011541115.2 XP_011539417.1 83 - 84 12318 88
XM_011541116.2 XP_011539418.1 82 - 83 12309 88
XM_011541117.2 XP_011539419.1 82 - 83 12258 88
XM_011541118.2 XP_011539420.1 82 - 83 12255 88
XM_011541119.2 XP_011539421.1 83 - 84 12342 88
XM_011541120.2 XP_011539422.1 82 - 83 12219 88
XM_011541121.2 XP_011539423.1 81 - 82 12186 88
XM_017000822.1 XP_016856311.1 83 - 84 12321 88
XM_017000823.1 XP_016856312.1 82 - 83 12294 88
XM_017000824.1 XP_016856313.1 82 - 83 12240 88
XM_017000825.1 XP_016856314.1 81 - 82 12225 88
XM_017000826.1 XP_016856315.1 82 - 83 12222 88
XM_017000827.1 XP_016856316.1 81 - 82 12207 88
XM_017000828.1 XP_016856317.1 81 - 82 12183 88
XM_017000829.1 XP_016856318.1 81 - 82 12135 88
XM_017000830.1 XP_016856319.1 80 - 81 12084 88
GenBank mRNA AB007931.1 - 22 - 23 3383 88
AF348492.1 - 81 - 82 12103 88
AK023826.1 - 19 - 20 2938 88

View Full Product Details