Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLRX3
Entrez Gene ID
10539
Gene Name
glutaredoxin 3
Gene Aliases
GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3
Chromosome Location
Chr.10: 130136375 - 130182877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199868.1 NP_001186797.1 8 - 9 868 155
NM_001321980.1 NP_001308909.1 9 - 10 923 155
NM_006541.4 NP_006532.2 8 - 9 868 155
XM_017015474.1 XP_016870963.1 9 - 10 923 155
GenBank mRNA AF118649.1 - 8 - 9 831 155
AF118652.1 - 8 - 9 834 155
AJ010841.1 - 8 - 9 826 155
AK022131.1 - 8 - 9 842 155
AK225609.1 - 8 - 9 831 155
BC005289.1 - 8 - 9 844 155
BC014372.1 - 7 - 8 686 155
BI256922.1 - 3 - 4 209 155

View Full Product Details