The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199868.1 | NP_001186797.1 | 9 - 10 | 914 | 125 |
NM_001321980.1 | NP_001308909.1 | 10 - 11 | 969 | 125 | |
NM_006541.4 | NP_006532.2 | 9 - 10 | 914 | 125 | |
XM_017015474.1 | XP_016870963.1 | 10 - 11 | 969 | 125 | |
GenBank mRNA | AF118649.1 | - | 9 - 10 | 877 | 125 |
AF118652.1 | - | 9 - 10 | 880 | 125 | |
AJ010841.1 | - | 9 - 10 | 872 | 125 | |
AK022131.1 | - | 9 - 10 | 888 | 125 | |
AK225609.1 | - | 9 - 10 | 877 | 125 | |
BC005289.1 | - | 9 - 10 | 890 | 125 | |
BC014372.1 | - | 8 - 9 | 732 | 125 | |
BI256922.1 | - | 4 - 5 | 255 | 125 |