The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153213.3 | NP_694945.2 | 6 - 7 | 1278 | 158 |
XM_011540706.2 | XP_011539008.1 | 7 - 8 | 1442 | 158 | |
GenBank mRNA | AK091281.1 | - | 6 - 7 | 1314 | 158 |
AK297707.1 | - | 6 - 7 | 1340 | 158 | |
BC012982.2 | - | 5 - 6 | 854 | 158 | |
BC040640.1 | - | 6 - 7 | 1278 | 158 |