Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBR4
Entrez Gene ID
23352
Gene Name
ubiquitin protein ligase E3 component n-recognin 4
Gene Aliases
RBAF600, ZUBR1, p600
Chromosome Location
Chr.1: 19074506 - 19210334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020765.2 NP_065816.2 59 - 60 8740 146
XM_011541108.2 XP_011539410.1 61 - 62 8975 146
XM_011541109.2 XP_011539411.1 61 - 62 8972 146
XM_011541110.2 XP_011539412.1 61 - 62 8972 146
XM_011541111.2 XP_011539413.1 61 - 62 8972 146
XM_011541112.2 XP_011539414.1 61 - 62 8960 146
XM_011541113.2 XP_011539415.1 61 - 62 8957 146
XM_011541114.2 XP_011539416.1 61 - 62 8957 146
XM_011541115.2 XP_011539417.1 61 - 62 8951 146
XM_011541116.2 XP_011539418.1 60 - 61 8942 146
XM_011541117.2 XP_011539419.1 60 - 61 8891 146
XM_011541118.2 XP_011539420.1 60 - 61 8888 146
XM_011541119.2 XP_011539421.1 61 - 62 8975 146
XM_011541120.2 XP_011539422.1 60 - 61 8852 146
XM_011541121.2 XP_011539423.1 59 - 60 8819 146
XM_017000822.1 XP_016856311.1 61 - 62 8954 146
XM_017000823.1 XP_016856312.1 60 - 61 8927 146
XM_017000824.1 XP_016856313.1 60 - 61 8873 146
XM_017000825.1 XP_016856314.1 59 - 60 8858 146
XM_017000826.1 XP_016856315.1 60 - 61 8855 146
XM_017000827.1 XP_016856316.1 59 - 60 8840 146
XM_017000828.1 XP_016856317.1 59 - 60 8816 146
XM_017000829.1 XP_016856318.1 59 - 60 8768 146
XM_017000830.1 XP_016856319.1 58 - 59 8717 146
GenBank mRNA AF348492.1 - 59 - 60 8736 146
AK000331.1 - 12 - 13 1560 146

View Full Product Details